Learn More
BOULE Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-13720-100UL
This item is not returnable.
View return policy
Description
The BOULE antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human BOULE. This antibody recognizes human, mouse, rat antigen. The BOULE antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
BOULE | |
Polyclonal | |
Rabbit | |
Primary | |
Human | |
IgG |
Immunofluorescence | |
CoraLite 488 | |
100 μL | |
66037 | |
Liquid |