missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boron oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific™
502.00 € - 1958.00 €
Chemical Identifiers
CAS | 1303-86-2 |
---|---|
Molecular Formula | B2O3 |
Molecular Weight (g/mol) | 69.62 |
MDL Number | MFCD00011315 |
InChI Key | JKWMSGQKBLHBQQ-UHFFFAOYSA-N |
Synonym | boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g |
PubChem CID | 518682 |
ChEBI | CHEBI:30163 |
IUPAC Name | oxo(oxoboranyloxy)borane |
SMILES | O=BOB=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11366178
|
Thermo Scientific Alfa Aesar
011158.22 |
100 g |
502.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11376178
|
Thermo Scientific Alfa Aesar
011158.36 |
500 g | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- In the electronics industry as a liquid encapsulent for Czochralski-type growth of III-V semiconductor single crystals (e.g., GaAs, GaP, InP)
Chemical Identifiers
1303-86-2 | |
69.62 | |
JKWMSGQKBLHBQQ-UHFFFAOYSA-N | |
518682 | |
oxo(oxoboranyloxy)borane |
B2O3 | |
MFCD00011315 | |
boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g | |
CHEBI:30163 | |
O=BOB=O |
Specifications
1860°C | |
1303-86-2 | |
Fused Lump | |
MFCD00011315 | |
boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g | |
O=BOB=O | |
69.62 | |
CHEBI:30163 | |
99.999% | |
Odorless | |
2.46 g/mL (±)0.01 | |
Boron oxide, Fused Lump, Approx. 1.0 in. dia. x 0.75 in. thick |
∼450°C | |
White | |
B2O3 | |
14,1337 | |
JKWMSGQKBLHBQQ-UHFFFAOYSA-N | |
oxo(oxoboranyloxy)borane | |
518682 | |
69.62 | |
Puratronic™ | |
Hygroscopic | |
(metals basis) |
Safety and Handling
GHS H Statement
H360
May damage fertility or the unborn child.
P201-P202-P281-P308+P313-P501c
H360FD
missing translation for 'einecsNumber' : 215-125-8
missing translation for 'rtecsNumber' : ED7900000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only