missing translation for 'onlineSavingsMsg'
Learn More

Boron oxide, 99.98% (metals basis), Thermo Scientific Chemicals

Product Code. 11493870
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Boron oxide is used as a flux, an acid catalyst and a chemical intermediate in the production of boron halides, esters, carbide, nitride and metallic borides. Boron oxide has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from talc. It is actively involved in the production of borosilicate glass. It is an additive used in high-temperature lubricants and dopants in semiconductor materials.Boron oxide is used as a fluxing agent for glass and enamels and as an additive used in glass fibers. It is used in the production of borosilicate glass, the raw material of chemical heat-treatment dissolved boron, organic synthesis catalyst and dopants use in semiconductor materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 1303-86-2
Molecular Formula B2O3
Molecular Weight (g/mol) 69.62
MDL Number MFCD00011315
InChI Key JKWMSGQKBLHBQQ-UHFFFAOYSA-N
Synonym boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g
PubChem CID 518682
ChEBI CHEBI:30163
IUPAC Name oxo[(oxoboranyl)oxy]borane
SMILES O=BOB=O

Spécification

Boiling Point 1860°C
Melting Point ∽450°C
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.98% (Metal Basis)
Solubility Information Soluble in water. Slightly soluble in methanol.
Formula Weight 69.62
Odor Odorless
Sensitivity Hygroscopic
Appearance White powder
Density 2.46 g/cm3;
Chemical Name or Material Boron oxide
Afficher plus Afficher moins

RUO – Research Use Only

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis