missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boron nitride, 99.5% (metals basis), Thermo Scientific Chemicals
192.00 € - 540.00 €
Chemical Identifiers
CAS | 10043-11-5 |
---|---|
Molecular Formula | BN |
Molecular Weight (g/mol) | 24.82 |
MDL Number | MFCD00011317 |
InChI Key | LNLSXDSWJBUPHM-UHFFFAOYSA-N |
Synonym | boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r |
PubChem CID | 66227 |
ChEBI | CHEBI:50883 |
SMILES | B=N |
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
15462067
|
Thermo Scientific Alfa Aesar
044917.KG |
1 Pc. |
192.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15472067
|
Thermo Scientific Alfa Aesar
044917.KJ |
5 Pc. | |||||||
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificadores de productos químicos
10043-11-5 | |
24.82 | |
LNLSXDSWJBUPHM-UHFFFAOYSA-N | |
66227 | |
B=N |
BN | |
MFCD00011317 | |
boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r | |
CHEBI:50883 |
Especificaciones
10043-11-5 | |
BN | |
boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r | |
B=N | |
24.82 | |
CHEBI:50883 | |
99.5% | |
Boron nitride |
Hot Pressed Rod | |
MFCD00011317 | |
LNLSXDSWJBUPHM-UHFFFAOYSA-N | |
azanylidyneborane | |
66227 | |
24.82 | |
Bag |
Seguridad y manipulación
P261-P271-P304+P340-P312-P501c
H335
missing translation for 'einecsNumber' : 233-136-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only