missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boron nitride, 99.5% (metals basis), Thermo Scientific™
Used in production of alloys. It is also used in the manufacture of furnace refractories, insulation materials, machinery and aviation.
203.00 € - 838.00 €
Chemical Identifiers
CAS | 10043-11-5 |
---|---|
Molecular Formula | BN |
Molecular Weight (g/mol) | 24.82 |
MDL Number | MFCD00011317 |
InChI Key | LNLSXDSWJBUPHM-UHFFFAOYSA-N |
Synonym | boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r |
PubChem CID | 66227 |
ChEBI | CHEBI:50883 |
IUPAC Name | boranylidyneamine |
SMILES | B=N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15412047
|
Thermo Scientific Alfa Aesar
044838.KG |
1 Pc. |
203.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15422047
|
Thermo Scientific Alfa Aesar
044838.KJ |
5 Pc. | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
10043-11-5 | |
24.82 | |
LNLSXDSWJBUPHM-UHFFFAOYSA-N | |
66227 | |
boranylidyneamine |
BN | |
MFCD00011317 | |
boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r | |
CHEBI:50883 | |
B=N |
Specifications
10043-11-5 | |
BN | |
boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r | |
LNLSXDSWJBUPHM-UHFFFAOYSA-N | |
boranylidyneamine | |
66227 | |
24.82 | |
Bag |
Hot Pressed Rod | |
MFCD00011317 | |
Insoluble in water. | |
B=N | |
24.82 | |
CHEBI:50883 | |
99.5% | |
Boron nitride |
Safety and Handling
P261-P271-P304+P340-P312-P501c
H335
missing translation for 'einecsNumber' : 233-136-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures