missing translation for 'onlineSavingsMsg'
Learn More

Boron nitride, 99.5% (metals basis), Thermo Scientific™

Used in production of alloys. It is also used in the manufacture of furnace refractories, insulation materials, machinery and aviation.

320.00 € - 1180.00 €

Chemical Identifiers

CAS 10043-11-5
Molecular Formula BN
Molecular Weight (g/mol) 24.82
MDL Number MFCD00011317
InChI Key LNLSXDSWJBUPHM-UHFFFAOYSA-N
Synonym boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r
PubChem CID 66227
ChEBI CHEBI:50883
IUPAC Name boranylidyneamine
SMILES B=N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15492037
View Documents
Thermo Scientific Alfa Aesar
044837.KG
1 Pc. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15402047
View Documents
Thermo Scientific Alfa Aesar
044837.KJ
5 Pc. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

10043-11-5
24.82
LNLSXDSWJBUPHM-UHFFFAOYSA-N
66227
boranylidyneamine
BN
MFCD00011317
boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r
CHEBI:50883
B=N

Specifications

10043-11-5
BN
boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r
LNLSXDSWJBUPHM-UHFFFAOYSA-N
boranylidyneamine
66227
24.82
Bag
Hot Pressed Rod
MFCD00011317
Insoluble in water.
B=N
24.82
CHEBI:50883
99.5%
Boron nitride
Videos
Safety and Handling

Safety and Handling

P261-P271-P304+P340-P312-P501c

H335

missing translation for 'einecsNumber' : 233-136-6

missing translation for 'rtecsNumber' : ED7800000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.