Learn More
BORIS Mouse anti-Human, CoraLite 488, Clone: 2B12E2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-60079-100UL
Product Code. 16810586
  • 450.00 € / 100µL
Explore more special offers
Description
Description
The BORIS antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human BORIS. This antibody recognizes human, mouse antigen. The BORIS antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
Specifications
BORIS
Monoclonal
2000 μg/mL
Mouse
Primary
Human, Mouse
Immunofluorescence
2B12E2
CoraLite 488
100 μL
140690
IgG1