Learn More
Bohlender™ Bola PTFE/PPS Ground Joint GL Adaptors
Assemble components with ground joints (condensers, stirrer bearings etc.) on glass bottles or GL-threaded necks with these adaptors. Bohlender™ BOLA™ Ground Joint GL Adaptors can be removed and fixed on another bottle without the risk of disarranging the tubing.
Brand:  Bohlender™ D734-40
Product Code. 10422943
  • 137.00 € / Each
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Black screw cap made of PPS with GL 45 thread
  • Movable insert with ground joint made of PTFE
  • Transition from a ground joint to a glass thread
  • The body can be turned independently from the screw cap
  • Very good chemical resistance
  • For working temperatures up to +250°C
Specifications
Specifications
Assembly of components with ground joint (condensers, stirrer bearings etc.) on glass bottles or GL-threaded necks.
Ground Joint GL Adaptors
29/32
PPS, PTFE
1 Each