Learn More
Helios Italquartz Boats with Ring Handle, Transparent
Boats with ring handle constructed of transparent, durable, resistant, and chemically inert quartz glass.
Brand:  Helios Italquartz 62P00009
 View more versions of this product
Product Code. 15888034
  • 255.00 € / Each
Estimated Shipment: 06-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Transparent
  • Include ring handle
  • Chemically inert
  • Excellent electrical insulation
  • Excellent strength and durability
  • Exceptional thermal stability compared to glass
Specifications
Specifications
Flat
Quartz
Boat with Ring Handle
143/33 mm
17 mm
Transparent
Boat
143 mm x 17 mm
17 mm