Learn More
Helios Italquartz Boats with Ring Handle, Transparent
Boats with ring handle constructed of transparent, durable, resistant, and chemically inert quartz glass.
Brand:  Helios Italquartz 62P00008
 View more versions of this product
Product Code. 15878034
  • 192.00 € / Each
Estimated Shipment: 06-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Transparent
  • Include ring handle
  • Chemically inert
  • Excellent electrical insulation
  • Excellent strength and durability
  • Exceptional thermal stability compared to glass
Specifications
Specifications
Transparent
Boat with Ring Handle
180/20 mm
Quartz
180 mm x 15 mm
15 mm