Learn More
Berkshire™ BlueSorb™ 750
BlueSorb™ 750 is absorbent with low extractable levels and particle counts. Excellent combination of the synthetic polyester strength and cleanliness with the absorbent characteristics of cellulose. Blue color allows for differentiation in critical areas.
Brand: Berkshire™ BS750.1212.20
This item is not returnable.
View return policy
Description
- Excellent combination of the synthetic polyester strength and cleanliness with the absorbent characteristics of cellulose
- Blue color allows differentiation for guarding against migration into critical areas
- Smooth, absorbent and durable
- Low extractable levels and particle counts
Specifications
Surface Wiper | |
30 cm | |
Polyester Cellulose | |
ISO Class 5-8 |
12 in. | |
30 cm | |
Blue |