Learn More
Sampling Systems™ Blue SteriWare™ Liquid Funnels
BIOPHARMA
Remove the risk of cross contamination and improve your quality control practices by dispensing liquids with blue SteriWare™ Liquid Funnels. Manufactured in our cleanroom and available sterile, these funnels are ideal for use in a wide range of applications in the food industry.
58.15 € - 139.00 €
Specifications
Diameter (Metric)Top 100 mm
Material Polypropylene
Color Blue
View More Specs
Products 2
Product Code Brand Sterility Price Quantity & Availability  
Product Code Brand Sterility Price Quantity & Availability  
17574656
BIOPHARMA
View Documents
Sampling Systems™
8370B-100
Non-sterile
58.15 €
Pack of 50
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
17584656
BIOPHARMA
View Documents
Sampling Systems™
8370B-100S
Sterile
139.00 €
Pack of 50
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
Description
Description
  • Our range of single use, disposable liquid funnels are ideal for use in laboratories and manufacturing areas.
  • These funnels are made from food grade polymers and are designed to be used straight from the bag.
  • The bright blue color means they can be easily seen if they drop in or left in the material by mistake.
  • SteriWare™ Funnels are available sterile and non-sterile.
  • SteriWare™ is the largest range of single-use equipment for sampling, laboratory and production work on the market.
  • The whole range is made in a cleanroom and comes with class-leading pharmaceutical documentation.
  • Simply unwrap the funnel and use.
  • The funnels come with a side tab so they are easy to hold.
  • The individually bagged funnels are made from PP that is FDA/EC compliant.
  • The material used is also certified BSE/TSE free. The sterile funnels conform to SAL 10-6
  • The funnels come with a batch-specific certificate that includes the date of manufacture, expiry date and irradiation certificate (for sterile funnels only).
Specifications
Specifications
100 mm
Polypropylene
Blue