missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BL21(DE3)-lysate Rabbit anti-Bacteria, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | BL21(DE3)-lysate |
---|---|
Concentration | 0.7 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16897013
|
Proteintech
10558-1-IG-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16887013
|
Proteintech
10558-1-IG-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Escherichia coli B serves as a research model for studying phage sensitivity, restriction-modification systems, and bacterial evolution, and also as a workhorse for protein expression in life science laboratories and in the biotech industry. Characteristics such as protease deficiency, low acetate production at a high level of glucose, and enhanced permeability (probably due to a simple cell surface) make E. coli B a desirable host for the production of genetically engineered proteins. BL21(DE3) has been very widely used to express recombit proteins. Comparison of BL21(DE3) and another B strain REL606 shows they differ in length by 72. 304 bp and have 426 single nucleotide polymorphisms, all of which have been accounted for by various laboratory manipulations. Differences between B strains and K12 include the absence of flagellar component genes, the DNA cytosine methylase dcm, and ompT in BL21(DE3). B strains may have an additional type II secretion system not found in K12. Other differences include differential distribution of insertions sequences and other changes resulting from horizontal gene transfer. BL21(DE3) also carries a DE3 recombit phage harboring the T7 RNA polymerase gene that can direct high-level expression of cloned genes under the control of the T7 promoter.Specifications
BL21(DE3)-lysate | |
Western Blot | |
Unconjugated | |
Rabbit | |
Bacteria | |
BL21(DE3) lysate | |
IgG | |
Protein A | |
Antibody |
0.7 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
BL21(DE3)-lysate Fusion Protein Ag0048 | |
Primary | |
-20°C |