missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth pieces, 10cm (3.9in) & down, 99.99% (metals basis), Thermo Scientific Chemicals
98.80 € - 886.00 €
Chemical Identifiers
CAS | 7440-69-9 |
---|---|
Molecular Formula | Bi |
Molecular Weight (g/mol) | 208.98 |
MDL Number | MFCD00134033 |
InChI Key | JCXGWMGPZLAOME-UHFFFAOYSA-N |
Synonym | bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod |
PubChem CID | 5359367 |
ChEBI | CHEBI:33301 |
IUPAC Name | bismuth |
SMILES | [Bi] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11316618
|
Thermo Scientific Alfa Aesar
012208.30 |
250 g |
98.80 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11306618
|
Thermo Scientific Alfa Aesar
012208.A1 |
1 kg |
220.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11326618
|
Thermo Scientific Alfa Aesar
012208.A7 |
5 kg |
886.00 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-69-9 | |
208.98 | |
JCXGWMGPZLAOME-UHFFFAOYSA-N | |
5359367 | |
bismuth |
Bi | |
MFCD00134033 | |
bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod | |
CHEBI:33301 | |
[Bi] |
Specifications
Gray | |
Gray needles | |
Bi | |
bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod | |
[Bi] | |
208.98 | |
CHEBI:33301 | |
1 hPa (1mm Hg) | |
Bismuth |
7440-69-9 | |
99.99% (Metals basis) | |
MFCD00134033 | |
JCXGWMGPZLAOME-UHFFFAOYSA-N | |
bismuth | |
5359367 | |
Odorless | |
Pieces |
Safety and Handling
missing translation for 'einecsNumber' : 231-177-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only