missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth(III) sulfide, 99%, pure, Thermo Scientific Chemicals
103.00 € - 635.00 €
Chemical Identifiers
CAS | 1345-07-9 |
---|---|
Molecular Formula | Bi2S3 |
Molecular Weight (g/mol) | 514.14 |
MDL Number | MFCD00014204 |
InChI Key | YNRGZHRFBQOYPP-UHFFFAOYSA-N |
IUPAC Name | dibismuth(3+) trisulfanediide |
SMILES | [S--].[S--].[S--].[Bi+3].[Bi+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15299926
|
Thermo Scientific Acros
453991000 |
100 g |
103.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15209936
|
Thermo Scientific Acros
453995000 |
500 g |
277.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15289926
|
Thermo Scientific Acros
453990025 |
2.5 kg | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
1345-07-9 | |
514.14 | |
YNRGZHRFBQOYPP-UHFFFAOYSA-N | |
[S--].[S--].[S--].[Bi+3].[Bi+3] |
Bi2S3 | |
MFCD00014204 | |
dibismuth(3+) trisulfanediide |
Specifications
1345-07-9 | |
Bi2S3 | |
MFCD00014204 | |
YNRGZHRFBQOYPP-UHFFFAOYSA-N | |
dibismuth(3+) trisulfanediide | |
514.16 | |
Pure |
80.1 to 82.5% (Bi) (Complexometry) | |
Bi2S3 | |
15,1287 | |
[S--].[S--].[S--].[Bi+3].[Bi+3] | |
514.14 | |
99% | |
Bismuth(III) sulfide |
Safety and Handling
missing translation for 'einecsNumber' : 215-716-
RUO – Research Use Only