Learn More
Bismuth(III) sulfide, 99.999% (metals basis), Thermo Scientific Chemicals
333.00 € - 1340.00 €
Chemical Identifiers
CAS | 1345-07-9 |
---|---|
Molecular Formula | Bi2S3 |
Molecular Weight (g/mol) | 514.14 |
MDL Number | MFCD00014204 |
InChI Key | YNRGZHRFBQOYPP-UHFFFAOYSA-N |
IUPAC Name | dibismuth(3+) trisulfanediide |
SMILES | [S--].[S--].[S--].[Bi+3].[Bi+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15442287
|
Thermo Scientific Alfa Aesar
045463.18 |
50 g |
333.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15452287
|
Thermo Scientific Alfa Aesar
045463.30 |
250 g |
1340.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bismuth(III) sulfide is mainly used in solar cells, photodiode array and infrared spectroscopy. It is also applied in the manufacture of other bismuth compounds, additives for free-cutting steels and micro-electronics industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1345-07-9 | |
514.14 | |
YNRGZHRFBQOYPP-UHFFFAOYSA-N | |
[S--].[S--].[S--].[Bi+3].[Bi+3] |
Bi2S3 | |
MFCD00014204 | |
dibismuth(3+) trisulfanediide |
Specifications
1345-07-9 | |
Lump | |
Bi2S3 | |
Insoluble in water. | |
[S--].[S--].[S--].[Bi+3].[Bi+3] | |
514.14 | |
7.7 g/cm3 |
Gray | |
99.999% (Metals basis) | |
MFCD00014204 | |
YNRGZHRFBQOYPP-UHFFFAOYSA-N | |
dibismuth(3+) trisulfanediide | |
514.15 | |
Bismuth(III) sulfide |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.
H315-H319-H335-H303: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May be harmful if swallowed.
missing translation for 'einecsNumber' : 215-716-0
missing translation for 'rtecsNumber' : EB3105000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Bismuth(III) sulfide
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P321-Specific treatment (see on this label).
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only