missing translation for 'onlineSavingsMsg'
Learn More
Bismuth(III) sulfate, 99%, pure, Thermo Scientific Chemicals Product Code.: 15209876

Bismuth(III) sulfate, 99%, pure, Thermo Scientific Chemicals

Product Code. 15209876
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 15209876

Brand: Thermo Scientific Acros 453850250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-68-0
Molecular Formula Bi2O12S3
Molecular Weight (g/mol) 706.13
MDL Number MFCD00036140
InChI Key BEQZMQXCOWIHRY-UHFFFAOYSA-H
Synonym bismuth sulfate, dibismuth trisulfate, bismuth trisulfate, sulfuric acid, bismuth 3+ salt 3:2, bismuth sulphate, bismuth sesquisulfate, bismuth sulfate mi, dibismuth tris sulphate, bismuth iii sulfate, 2bi.3so4Show MoreShow Less
PubChem CID 198144
IUPAC Name dibismuth;trisulfate
SMILES [Bi+3].[Bi+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Melting Point 405.0°C
Quantity 25 g
Assay Percent Range 98.5% min
Linear Formula Bi2(SO4)3
Merck Index 15,1286
Formula Weight 706.13
Percent Purity 99%
Grade Pure
Chemical Name or Material Bismuth(III) sulfate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0395768 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number453851000,453850250
Lot NumberA0396300 Certificate TypeCertificate of Analysis Date05/11/2020 Catalog Number
Lot NumberA0391403 Certificate TypeCertificate of Analysis Date10/06/2020 Catalog Number453851000,453850250
Product Identifier
  • Bismuth(III) sulfate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bismuth(III) sulfate, 99%, pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.