Learn More
Bismuth(III) oxide, nanopowder, 99.9%, Thermo Scientific Chemicals
33.90 € - 405.00 €
Chemical Identifiers
CAS | 1304-76-3 |
---|---|
Molecular Formula | Bi2O3 |
Molecular Weight (g/mol) | 465.96 |
MDL Number | MFCD00003462 |
InChI Key | TYIXMATWDRGMPF-UHFFFAOYSA-N |
IUPAC Name | dibismuth(3+) trioxidandiide |
SMILES | [O--].[O--].[O--].[Bi+3].[Bi+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15402657
|
Thermo Scientific Alfa Aesar
046314.14 |
25 g |
33.90 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15412657
|
Thermo Scientific Alfa Aesar
046314.22 |
100 g |
98.70 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15422657
|
Thermo Scientific Alfa Aesar
046314.36 |
500 g |
405.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bismuth(III) oxide is used in the preparation of BiFeO3perovskite nanoparticles. It finds use in disinfectants, magnets, glass, rubber, vulcanization, fireproofing papers and polymers and catalysts. Bismuth trioxide brings about the dragon's eggs effect in fireworks, as a replacement of red lead. Bismuth(III) compounds are attractive reagents and catalysts in organic synthesis because of their low cost and ease of handling. Bismuth trioxide nanoparticles also play an important role in high energy gas generators. The alpha crystalline form of bismuth(III) oxide has p-type electronic conductivity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1304-76-3 | |
465.96 | |
TYIXMATWDRGMPF-UHFFFAOYSA-N | |
[O--].[O--].[O--].[Bi+3].[Bi+3] |
Bi2O3 | |
MFCD00003462 | |
dibismuth(3+) trioxidandiide |
Specifications
∼1890°C | |
1304-76-3 | |
Powder | |
MFCD00003462 | |
Soluble in acids. Insoluble in water. | |
[O--].[O--].[O--].[Bi+3].[Bi+3] | |
465.96 | |
99.9% | |
Bismuth (III) oxide |
817°C | |
Yellow | |
Bi2O3 | |
14,1273 | |
TYIXMATWDRGMPF-UHFFFAOYSA-N | |
dibismuth(3+) trioxidandiide | |
465.96 | |
8.9 g/mL |
Safety and Handling
missing translation for 'einecsNumber' : 215-134-7
missing translation for 'rtecsNumber' : EB2984460
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only