missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) oxide, NanoArc™, 99.5+%, Thermo Scientific Chemicals

40.20 € - 415.00 €

Chemical Identifiers

CAS 1304-76-3
Molecular Formula Bi2O3
Molecular Weight (g/mol) 465.96
MDL Number MFCD00003462
InChI Key TYIXMATWDRGMPF-UHFFFAOYSA-N
IUPAC Name dibismuth(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Bi+3].[Bi+3]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15422387
View Documents
Thermo Scientific Alfa Aesar
045582.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15432387
View Documents
Thermo Scientific Alfa Aesar
045582.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15442387
View Documents
Thermo Scientific Alfa Aesar
045582.36
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Bismuth(III) oxide is used in solid-oxide fuel cells. It is also used in optical glasses, to replace lead oxide in whitewares, luxes, varistor formulations and ceramic capacitor formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • X-ray opaque particle
  • NanoArc™ bismuth oxide can be compounded into polymers or coating systems to increase opacity to x-rays,while the small particle size ensures minimal disruption of physical properties
Specifications

Chemical Identifiers

1304-76-3
465.96
TYIXMATWDRGMPF-UHFFFAOYSA-N
[O--].[O--].[O--].[Bi+3].[Bi+3]
Bi2O3
MFCD00003462
dibismuth(3+) trioxidandiide

Specifications

∼1890°C
1304-76-3
Powder
MFCD00003462
Soluble in acids. Insoluble in water.
[O--].[O--].[O--].[Bi+3].[Bi+3]
465.96
≥99.5%
8.9 g/mL
817°C
Yellow
Bi2O3
14,1273
TYIXMATWDRGMPF-UHFFFAOYSA-N
dibismuth(3+) trioxidandiide
465.96
Odorless
Bismuth (III) oxide NanoArc™ BI-7300
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-134-7

missing translation for 'rtecsNumber' : EB2984460

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.