Learn More
Bismuth(III) oxide, 99.975% (metals basis), Thermo Scientific Chemicals
62.30 € - 347.00 €
Chemical Identifiers
CAS | 1304-76-3 |
---|---|
Molecular Formula | Bi2O3 |
Molecular Weight (g/mol) | 465.96 |
MDL Number | MFCD00003462 |
InChI Key | TYIXMATWDRGMPF-UHFFFAOYSA-N |
Synonym | Bismuth salicylate, basic, Salicylic acid bismuth basic salt |
IUPAC Name | dibismuth(3+) trioxidandiide |
SMILES | [O--].[O--].[O--].[Bi+3].[Bi+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11363978
|
Thermo Scientific Alfa Aesar
040394.18 |
50 g |
62.30 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11353978
|
Thermo Scientific Alfa Aesar
040394.30 |
250 g |
154.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11373978
|
Thermo Scientific Alfa Aesar
040394.A1 |
1 kg |
347.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used as Adhesives and sealant chemicals, flame retardants, pigments, propellants and blowing agents. It is also used as solvents. Other uses are in batteries, electrical and electronic products, explosive materials ink, toner, and colorant products, paints and coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1304-76-3 | |
465.96 | |
TYIXMATWDRGMPF-UHFFFAOYSA-N | |
dibismuth(3+) trioxidandiide |
Bi2O3 | |
MFCD00003462 | |
Bismuth salicylate, basic, Salicylic acid bismuth basic salt | |
[O--].[O--].[O--].[Bi+3].[Bi+3] |
Specifications
ca 1890°C | |
1304-76-3 | |
Powder | |
Bi2O3 | |
Bismuth salicylate, basic, Salicylic acid bismuth basic salt | |
TYIXMATWDRGMPF-UHFFFAOYSA-N | |
dibismuth(3+) trioxidandiide | |
465.96 | |
8.9 |
817°C | |
Yellow | |
99.975% (Metals basis) | |
MFCD00003462 | |
Insoluble in water. Soluble in acids. | |
[O--].[O--].[O--].[Bi+3].[Bi+3] | |
465.96 | |
Odorless | |
Bismuth(III) oxide |
Safety and Handling
H303: May be harmful if swallowed.
missing translation for 'einecsNumber' : 215-134-7
missing translation for 'rtecsNumber' : EB2984460
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only