missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) nitrate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

99.70 € - 320.60 €

Chemical Identifiers

CAS 10361-44-1
Molecular Formula BiN3O9
Molecular Weight (g/mol) 394.99
MDL Number MFCD00003460
InChI Key PPNKDDZCLDMRHS-UHFFFAOYSA-N
Synonym bismuth nitrate, bismuth trinitrate, ccris 7110, nitric acid, bismuth 3+ salt, tris nitrooxy bismuthine, bismuth 3+ trinitrate, bi.3no3, bismuth iii nitrate, bismuth iii nitrate hydrate, nitric acid, bismuth 3+ salt 3:1
PubChem CID 107711
IUPAC Name bismuth;trinitrate
SMILES [Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11380869
View Documents
Thermo Scientific Alfa Aesar
010657.14
25 g
99.70 €
25g
Please to purchase this item. Need a web account? Register with us today!
11370869
View Documents
Thermo Scientific Alfa Aesar
010657.22
100 g
320.60 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used in electronic ceramics and fluorescent paints. It is employed as an oxidant for selective oxidation of sulfides to sulfoxides, as a nitrating reagent for synthesizing nitro aromatic compounds, and as a precursor for Dragendorff reagent for use in TLC stain and for testing the presence of alkaloids. On silica gel support, it has been reported to be an efficient catalyst for the Biginelli reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10361-44-1
394.99
PPNKDDZCLDMRHS-UHFFFAOYSA-N
107711
[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
BiN3O9
MFCD00003460
bismuth nitrate, bismuth trinitrate, ccris 7110, nitric acid, bismuth 3+ salt, tris nitrooxy bismuthine, bismuth 3+ trinitrate, bi.3no3, bismuth iii nitrate, bismuth iii nitrate hydrate, nitric acid, bismuth 3+ salt 3:1
bismuth;trinitrate

Specifications

30°C (decomposition)
Colorless to White
BiN3O9
UN1477
bismuth nitrate, bismuth trinitrate, ccris 7110, nitric acid, bismuth 3+ salt, tris nitrooxy bismuthine, bismuth 3+ trinitrate, bi.3no3, bismuth iii nitrate, bismuth iii nitrate hydrate, nitric acid, bismuth 3+ salt 3:1
PPNKDDZCLDMRHS-UHFFFAOYSA-N
bismuth;trinitrate
107711
99.999%
Hygroscopic
Bismuth(III) nitrate hydrate
10361-44-1
Crystalline
MFCD00003460
14,1271
Slightly soluble in water; soluble in acetone,acetic acid,and glycerol. Insoluble in ethyl acetate and ethanol.
[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
394.99
394.99 (Anhydrous)
Puratronic™
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H272-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 233-791-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Bismuth(III) nitrate hydrate, Puratronic

Signal Word
  • Danger

Hazard Category
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.