Learn More
Bismuth(III) chloride oxide, 98%, Thermo Scientific Chemicals
Used as pigment, in manufacture of artificial pearls, dry-cell cathodes
51.30 € - 722.00 €
Chemical Identifiers
CAS | 7787-59-9 |
---|---|
Molecular Formula | BiClHO |
Molecular Weight (g/mol) | 261.437 |
MDL Number | MFCD00010895 |
InChI Key | BWOROQSFKKODDR-UHFFFAOYSA-N |
Synonym | bismuth oxychloride, mearlite, pearl white, bismuthine, chlorooxo, wismutchloridoxid, bismut-chlorid-oxid, biju, bismuth chloride oxide, basic bismuth chloride, wismut iii-chlorid-oxid |
PubChem CID | 6328152 |
IUPAC Name | oxobismuth;hydrochloride |
SMILES | O=[Bi].Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11303988
|
Thermo Scientific Alfa Aesar
040396.18 |
50 g |
51.30 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11323988
|
Thermo Scientific Alfa Aesar
040396.30 |
250 g |
214.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11313988
|
Thermo Scientific Alfa Aesar
040396.A1 |
1 kg |
722.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bismuth(III) chloride oxide is used in pharmaceutical industry, in the production of cosmetics, in chemical and metallurgical industries. It is used as pigment, in manufacture of artificial pearls and as a dry-cell cathodes, in photocatalysts for degradation of organic pollutant, in white-light LEDs, in the manufacture of plastics, in the synthesis of methanol, in semiconductor devices such as CCDs or photodiodes and also used in the production of permanent magnets.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7787-59-9 | |
261.437 | |
BWOROQSFKKODDR-UHFFFAOYSA-N | |
6328152 | |
O=[Bi].Cl |
BiClHO | |
MFCD00010895 | |
bismuth oxychloride, mearlite, pearl white, bismuthine, chlorooxo, wismutchloridoxid, bismut-chlorid-oxid, biju, bismuth chloride oxide, basic bismuth chloride, wismut iii-chlorid-oxid | |
oxobismuth;hydrochloride |
Specifications
(decomposition) | |
Brown | |
BiClHO | |
MFCD00010895 | |
bismuth oxychloride, mearlite, pearl white, bismuthine, chlorooxo, wismutchloridoxid, bismut-chlorid-oxid, biju, bismuth chloride oxide, basic bismuth chloride, wismut iii-chlorid-oxid | |
BWOROQSFKKODDR-UHFFFAOYSA-N | |
oxobismuth;hydrochloride | |
6328152 | |
98% | |
2.15 | |
Bismuth(III) chloride oxide |
7787-59-9 | |
Powder | |
BiOCl | |
14,1263 | |
Insoluble in water,alcohol; Soluble in acids. | |
O=[Bi].Cl | |
261.437 | |
260.43 | |
Odorless | |
7.72 g/mL |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 232-122-7
missing translation for 'rtecsNumber' : EB2700000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Bismuth(III) chloride oxide
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only