missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth(III) acetate, 99.999% (metals basis), Thermo Scientific Chemicals
145.00 € - 467.00 €
Chemical Identifiers
CAS | 22306-37-2 |
---|---|
Molecular Formula | C6H9BiO6 |
Molecular Weight (g/mol) | 386.112 |
MDL Number | MFCD00015629 |
InChI Key | WKLWZEWIYUTZNJ-UHFFFAOYSA-K |
Synonym | bismuth acetate, bismuth triacetate, unii-8aja86y692, acetic acid, bismuth 3+ salt, acmc-20allk, bismuth iii ; acetate, bismuth 3+ triacetate, trisacetic acid bismuth salt, bismuth 3+ ion tris acetate ion, bismuth iii acetate 50g |
PubChem CID | 31132 |
IUPAC Name | bismuth;triacetate |
SMILES | CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Bi+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11311918
|
Thermo Scientific Alfa Aesar
011846.09 |
10 g |
145.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11301918
|
Thermo Scientific Alfa Aesar
011846.18 |
50 g |
467.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used in biology, printing, in photographic films, textiles, and also in proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
22306-37-2 | |
386.112 | |
WKLWZEWIYUTZNJ-UHFFFAOYSA-K | |
31132 | |
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Bi+3] |
C6H9BiO6 | |
MFCD00015629 | |
bismuth acetate, bismuth triacetate, unii-8aja86y692, acetic acid, bismuth 3+ salt, acmc-20allk, bismuth iii ; acetate, bismuth 3+ triacetate, trisacetic acid bismuth salt, bismuth 3+ ion tris acetate ion, bismuth iii acetate 50g | |
bismuth;triacetate |
Specifications
(decomposition) | |
White | |
99.99% (Metals basis) | |
MFCD00015629 | |
Soluble in water. | |
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Bi+3] | |
386.112 | |
386.12 | |
Bismuth(III) acetate |
22306-37-2 | |
Crystals | |
C6H9BiO6 | |
bismuth acetate, bismuth triacetate, unii-8aja86y692, acetic acid, bismuth 3+ salt, acmc-20allk, bismuth iii ; acetate, bismuth 3+ triacetate, trisacetic acid bismuth salt, bismuth 3+ ion tris acetate ion, bismuth iii acetate 50g | |
WKLWZEWIYUTZNJ-UHFFFAOYSA-K | |
bismuth;triacetate | |
31132 | |
Hygroscopic |
Safety and Handling
missing translation for 'einecsNumber' : 244-904-5
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only