Learn More
Bismuth antimonide, 99.99%, Thermo Scientific Chemicals
68.00 € - 258.00 €
Chemical Identifiers
CAS | 12323-19-2 |
---|---|
Molecular Formula | Bi2Sb2 |
Molecular Weight (g/mol) | 661.48 |
MDL Number | MFCD00014196 |
InChI Key | AEMQIQQWIVNHAU-UHFFFAOYSA-N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11478866
|
Thermo Scientific Alfa Aesar
A17014.09 |
10 g |
68.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11488866
|
Thermo Scientific Alfa Aesar
A17014.18 |
50 g |
258.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12323-19-2 | |
661.48 | |
AEMQIQQWIVNHAU-UHFFFAOYSA-N |
Bi2Sb2 | |
MFCD00014196 |
Specifications
12323-19-2 | |
MFCD00014196 | |
AEMQIQQWIVNHAU-UHFFFAOYSA-N | |
330.73 | |
Bismuth antimonide |
Bi2Sb2 | |
UN1549 | |
661.48 | |
99.99% |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
- Bismuth antimonide
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P501-Dispose of contents/container to .
RUO – Research Use Only