missing translation for 'onlineSavingsMsg'
Learn More

Bis(trifluoromethanesulfonyl)imide, 99%, Thermo Scientific Chemicals

49.04 € - 509.54 €

Chemical Identifiers

CAS 82113-65-3
Molecular Formula C2HF6NO4S2
Molecular Weight (g/mol) 281.16
MDL Number MFCD00214154
InChI Key ZXMGHDIOOHOAAE-UHFFFAOYSA-N
Synonym 1,1,1-trifluoro-n-trifluoromethyl sulfonyl methanesulfonamide, trifluoromethanesulfonimide, unii-c7r849vm9l, bis trifluoromethanesulfonyl imide, bistrifluoromethanesulfonimide, 1,1,1-trifluoro-n-trifluoromethanesulfonylmethanesulfonamide, 1,1,1-trifluoro-n-trifluoromethylsulfonyl methanesulfonamide, bis trifluoromethane sulfonimide, n,n-bis trifluoromethanesulfonyl imide, n,n-bis trifluoromethylsulphonyl amine
PubChem CID 157857
IUPAC Name 1,1,1-trifluoro-N-(trifluoromethylsulfonyl)methanesulfonamide
SMILES C(F)(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10712024
View Documents Promotion Details
Thermo Scientific Acros
387600010
1 g Glass bottle
49.04 €
1g
Please to purchase this item. Need a web account? Register with us today!
10041593
View Documents Promotion Details
Thermo Scientific Acros
387600050
5 g Glass bottle
157.79 €
5g
Please to purchase this item. Need a web account? Register with us today!
10629004
View Documents Promotion Details
Thermo Scientific Acros
387600250
25 g Glass bottle
509.54 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
Specifications

Chemical Identifiers

82113-65-3
281.16
ZXMGHDIOOHOAAE-UHFFFAOYSA-N
157857
C(F)(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F
C2HF6NO4S2
MFCD00214154
1,1,1-trifluoro-n-trifluoromethyl sulfonyl methanesulfonamide, trifluoromethanesulfonimide, unii-c7r849vm9l, bis trifluoromethanesulfonyl imide, bistrifluoromethanesulfonimide, 1,1,1-trifluoro-n-trifluoromethanesulfonylmethanesulfonamide, 1,1,1-trifluoro-n-trifluoromethylsulfonyl methanesulfonamide, bis trifluoromethane sulfonimide, n,n-bis trifluoromethanesulfonyl imide, n,n-bis trifluoromethylsulphonyl amine
1,1,1-trifluoro-N-(trifluoromethylsulfonyl)methanesulfonamide

Specifications

82113-65-3
Brown to Colorless
Authentic
C2HF6NO4S2
1,1,1-trifluoro-n-trifluoromethyl sulfonyl methanesulfonamide, trifluoromethanesulfonimide, unii-c7r849vm9l, bis trifluoromethanesulfonyl imide, bistrifluoromethanesulfonimide, 1,1,1-trifluoro-n-trifluoromethanesulfonylmethanesulfonamide, 1,1,1-trifluoro-n-trifluoromethylsulfonyl methanesulfonamide, bis trifluoromethane sulfonimide, n,n-bis trifluoromethanesulfonyl imide, n,n-bis trifluoromethylsulphonyl amine
ZXMGHDIOOHOAAE-UHFFFAOYSA-N
1,1,1-trifluoro-N-(trifluoromethylsulfonyl)methanesulfonamide
157857
99%
Bis(trifluoromethanesulfonyl)imide
52°C to 56°C
90°C to 91°C
99%
MFCD00214154
Solubility in water: reacts
C(F)(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F
281.16
281.16
Crystals
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

Product Identifier
  • Bis(trifluoromethanesulfonyl)imide

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only