missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(phenylsulfonyl)methane, 99%, Thermo Scientific™
Click to view available options
Quantity:
25 g
Specifications
Specifications
CAS | 3406-02-8 |
Melting Point | 121.0°C to 122.0°C |
Flash Point | >110°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Molecular Formula | C13H12O4S2 |
Linear Formula | (C6H5SO2)2CH2 |
Quantity | 25 g |
Synonym | 2-phenylsulfonyl acetophenone, 2-phenylsulfonylacetophenone, 1-phenyl-2-phenylsulfonyl ethanone, acetophenone, 2-phenylsulfonyl, 2-benzenesulfonyl-1-phenylethanone, ethanone, 1-phenyl-2-phenylsulfonyl, 1-phenyl-2-phenylsulphonyl ethan-1-one, alpha-phenylsulfonylacetophenone, 2-benzenesulfonyl-1-phenylethan-1-one, 1-phenyl-2-phenylsulfonyl ethan-1-one |
InChI Key | DREVPGKOIZVPQV-UHFFFAOYSA-N |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'