Learn More
Bis(dichlorophosphino)methane, 96%, Thermo Scientific Chemicals
73.10 € - 259.00 €
Chemical Identifiers
CAS | 28240-68-8 |
---|---|
Molecular Formula | CH2Cl4P2 |
Molecular Weight (g/mol) | 217.79 |
MDL Number | MFCD01073767 |
InChI Key | OEKLCAXLANZKPX-UHFFFAOYSA-N |
Synonym | bis dichlorophosphino methane, dichloro dichlorophosphanyl methyl phosphane, acmc-1cq46, methylenebis dichlorophosphine, methylenebis phosphonous dichloride, dichloro dichlorophosphanylmethyl phosphane, phosphonous dichloride,p,p'-methylenebis |
PubChem CID | 2784185 |
IUPAC Name | dichloro(dichlorophosphanylmethyl)phosphane |
SMILES | C(P(Cl)Cl)P(Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10494505
|
Thermo Scientific Acros
383000010 |
1 g | Glass bottle |
73.10 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10228903
|
Thermo Scientific Acros
383000050 |
5 g | Glass bottle |
259.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
28240-68-8 | |
217.79 | |
OEKLCAXLANZKPX-UHFFFAOYSA-N | |
2784185 | |
C(P(Cl)Cl)P(Cl)Cl |
CH2Cl4P2 | |
MFCD01073767 | |
bis dichlorophosphino methane, dichloro dichlorophosphanyl methyl phosphane, acmc-1cq46, methylenebis dichlorophosphine, methylenebis phosphonous dichloride, dichloro dichlorophosphanylmethyl phosphane, phosphonous dichloride,p,p'-methylenebis | |
dichloro(dichlorophosphanylmethyl)phosphane |
Specifications
28240-68-8 | |
100.0 | |
1.6010g/mL | |
96°C | |
CH2Cl4P2 | |
1.5920 to 1.5950 | |
1.601 | |
Solubility in water: reacts | |
C(P(Cl)Cl)P(Cl)Cl | |
217.79 | |
217.79 | |
Liquid |
95.0 | |
Colorless to Yellow | |
101.0°C to 105.0°C (22.0 mmHg) | |
95% min. (Argentometry) | |
(Cl2P)2CH2 | |
MFCD01073767 | |
bis dichlorophosphino methane, dichloro dichlorophosphanyl methyl phosphane, acmc-1cq46, methylenebis dichlorophosphine, methylenebis phosphonous dichloride, dichloro dichlorophosphanylmethyl phosphane, phosphonous dichloride,p,p'-methylenebis | |
OEKLCAXLANZKPX-UHFFFAOYSA-N | |
dichloro(dichlorophosphanylmethyl)phosphane | |
2784185 | |
96% | |
Bis(dichlorophosphino)methane |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
- Bis(dichlorophosphino)methane
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only