Learn More
Bis(acetonitrile)(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, 97+%, Thermo Scientific Chemicals
687.00 € - 2947.00 €
Chemical Identifiers
CAS | 32679-02-0 |
---|---|
Molecular Formula | C12H18BF4N2Rh |
Molecular Weight (g/mol) | 380.00 |
MDL Number | MFCD06798085 |
InChI Key | KSMAFOANQHYEJG-SUESZSCISA-N |
Synonym | bis acetonitrile 1,5-cyclooctadiene rhodium 1 tetrafluoroborate, bis acetonitrile 1,5-cyclooctadiene rhodium i tetrafluoroborate, rh cod mecn 2 bf4, bis acetonitrile 1,5-cyclooctadiene rhodium i te |
PubChem CID | 11132655 |
SMILES | [Rh+].CC#N.CC#N.F[B-](F)(F)F.C1C\C=C/CC\C=C/1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15400097
|
Thermo Scientific Alfa Aesar
H66191.03 |
1 g |
687.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15410097
|
Thermo Scientific Alfa Aesar
H66191.06 |
5 g |
2947.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
32679-02-0 | |
380.00 | |
KSMAFOANQHYEJG-SUESZSCISA-N | |
11132655 |
C12H18BF4N2Rh | |
MFCD06798085 | |
bis acetonitrile 1,5-cyclooctadiene rhodium 1 tetrafluoroborate, bis acetonitrile 1,5-cyclooctadiene rhodium i tetrafluoroborate, rh cod mecn 2 bf4, bis acetonitrile 1,5-cyclooctadiene rhodium i te | |
[Rh+].CC#N.CC#N.F[B-](F)(F)F.C1C\C=C/CC\C=C/1 |
Specifications
32679-02-0 | |
C12H18BF4N2Rh | |
Air Sensitive | |
KSMAFOANQHYEJG-SUESZSCISA-N | |
bis(acetonitrile) cycloocta-1,5-diene tetrafluoroboranuide rhodium | |
11132655 | |
≥97% | |
Bis(acetonitrile)(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate |
Yellow | |
MFCD06798085 | |
bis acetonitrile 1,5-cyclooctadiene rhodium 1 tetrafluoroborate, bis acetonitrile 1,5-cyclooctadiene rhodium i tetrafluoroborate, rh cod mecn 2 bf4, bis acetonitrile 1,5-cyclooctadiene rhodium i te | |
[Rh+].CC#N.CC#N.F[B-](F)(F)F.C1C\C=C/CC\C=C/1 | |
380.00 | |
379.99 | |
Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only