missing translation for 'onlineSavingsMsg'
Learn More

Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, Thermo Scientific Chemicals

664.31 € - 2880.33 €

Chemical Identifiers

CAS 35138-22-8
Molecular Formula C16H24BF4Rh
Molecular Weight (g/mol) 406.08
MDL Number MFCD00075045
InChI Key NBGSCPNNKGVTKU-QMDOQEJBSA-N
Synonym bis 1,5-cyclooctadiene rhodium i tetrafluoroborate, rhodium i tetrafluoroborate 1,5-cyclooctadiene complex, 1z,5z-cycloocta-1,5-diene; rhodium; tetrafluoroborate, bis 1,5-cyclooctadiene rhodium tetrafluoroborate, rh cod 2 bf4, c16h24bf4rh, c16h24rh.bf4, bis 1,5-cyclooctadien rhodium i tetrafluoroborate
PubChem CID 5702662
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene;rhodium;tetrafluoroborate
SMILES [Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11430650
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044031.03
1 g
664.31 €
1g
Please to purchase this item. Need a web account? Register with us today!
11440650
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044031.06
5 g
2880.33 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate is useful for hydrogenation, isomerization reactions and is also employed as a precursor for asymmetric hydrogenations. Furthermore, it is used in the synthesis of cationic COD-Rhodium complexes with phosphine ligands. In addition, it shows an important application for enantioselective hydrogenation and hydrosilylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

35138-22-8
406.08
NBGSCPNNKGVTKU-QMDOQEJBSA-N
5702662
[Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
C16H24BF4Rh
MFCD00075045
bis 1,5-cyclooctadiene rhodium i tetrafluoroborate, rhodium i tetrafluoroborate 1,5-cyclooctadiene complex, 1z,5z-cycloocta-1,5-diene; rhodium; tetrafluoroborate, bis 1,5-cyclooctadiene rhodium tetrafluoroborate, rh cod 2 bf4, c16h24bf4rh, c16h24rh.bf4, bis 1,5-cyclooctadien rhodium i tetrafluoroborate
(1Z,5Z)-cycloocta-1,5-diene;rhodium;tetrafluoroborate

Specifications

35138-22-8
MFCD00075045
Insoluble in water.
[Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
406.08
406.08
Bis (1,5-cyclooctadiene)rhodium (I) tetrafluoroborate
C16H24BF4Rh
bis 1,5-cyclooctadiene rhodium i tetrafluoroborate, rhodium i tetrafluoroborate 1,5-cyclooctadiene complex, 1z,5z-cycloocta-1,5-diene; rhodium; tetrafluoroborate, bis 1,5-cyclooctadiene rhodium tetrafluoroborate, rh cod 2 bf4, c16h24bf4rh, c16h24rh.bf4, bis 1,5-cyclooctadien rhodium i tetrafluoroborate
NBGSCPNNKGVTKU-QMDOQEJBSA-N
(1Z,5Z)-cycloocta-1,5-diene;rhodium;tetrafluoroborate
5702662
Powder
Videos
Safety and Handling

Safety and Handling

P210-P234-P240-P241-P280-P370+P378q-P390

H228-H290-H500

missing translation for 'dotInformation' : Hazard Class: 4.1; Packaging Group: II

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only