missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(1,5-cyclooctadiene)iridium(I) tetrafluoroborate, Thermo Scientific Chemicals
190.00 € - 322.00 €
Chemical Identifiers
CAS | 35138-23-9 |
---|---|
Molecular Formula | C16H24BF4Ir |
Molecular Weight (g/mol) | 495.39 |
MDL Number | MFCD00079664 |
InChI Key | NKIQXDAGMKBVPE-QMDOQEJBSA-N |
Synonym | bis 1,5-cyclooctadiene iridium i tetrafluoroborate, ir cod 2 bf4, bis cyclooctadiene iridium i tetrafluoroborate, bis 1,5-cyclooctadiene iridium centn tetrafluoroborate, 1z,5z-cycloocta-1,5-diene; iridium; tetrafluoroborate, iridium 1+ ion bis 1,5-cyclooctadiene tetrafluoroborate, iridium 1+ bis 1,5-cyclooctadiene, z,z-tetrafluoroborate |
PubChem CID | 5702646 |
IUPAC Name | (1Z,5Z)-cycloocta-1,5-diene;iridium;tetrafluoroborate |
SMILES | [Ir+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11358198
|
Thermo Scientific Alfa Aesar
039414.ME |
500 mg |
190.00 €
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11368198
|
Thermo Scientific Alfa Aesar
039414.03 |
1 g |
322.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bis(1,5-cyclooctadiene)iridium(I) tetrafluoroborate is used as a halogenated compound for proteomics research. It is also used as intermediates in pharmaceuticals and chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
35138-23-9 | |
495.39 | |
NKIQXDAGMKBVPE-QMDOQEJBSA-N | |
5702646 | |
[Ir+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1 |
C16H24BF4Ir | |
MFCD00079664 | |
bis 1,5-cyclooctadiene iridium i tetrafluoroborate, ir cod 2 bf4, bis cyclooctadiene iridium i tetrafluoroborate, bis 1,5-cyclooctadiene iridium centn tetrafluoroborate, 1z,5z-cycloocta-1,5-diene; iridium; tetrafluoroborate, iridium 1+ ion bis 1,5-cyclooctadiene tetrafluoroborate, iridium 1+ bis 1,5-cyclooctadiene, z,z-tetrafluoroborate | |
(1Z,5Z)-cycloocta-1,5-diene;iridium;tetrafluoroborate |
Specifications
35138-23-9 | |
MFCD00079664 | |
Soluble in chloroform, methylene chloride | |
[Ir+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1 | |
495.39 | |
495.4 | |
Bis(1,5-cyclooctadiene)iridium(I) tetrafluoroborate |
C16H24BF4Ir | |
bis 1,5-cyclooctadiene iridium i tetrafluoroborate, ir cod 2 bf4, bis cyclooctadiene iridium i tetrafluoroborate, bis 1,5-cyclooctadiene iridium centn tetrafluoroborate, 1z,5z-cycloocta-1,5-diene; iridium; tetrafluoroborate, iridium 1+ ion bis 1,5-cyclooctadiene tetrafluoroborate, iridium 1+ bis 1,5-cyclooctadiene, z,z-tetrafluoroborate | |
NKIQXDAGMKBVPE-QMDOQEJBSA-N | |
(1Z,5Z)-cycloocta-1,5-diene;iridium;tetrafluoroborate | |
5702646 | |
Powder |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only