Learn More
Biphenyl-4-sulfonyl chloride, 97%, Thermo Scientific Chemicals
59.50 € - 690.00 €
Chemical Identifiers
CAS | 1623-93-4 |
---|---|
Molecular Formula | C12H9ClO2S |
Molecular Weight (g/mol) | 252.71 |
MDL Number | MFCD00078658 |
InChI Key | ALBQXDHCMLLQMB-UHFFFAOYSA-N |
Synonym | 4-biphenylsulfonyl chloride, 1,1'-biphenyl-4-sulfonyl chloride, biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulphonyl chloride, 4-biphenylsulfonylchloride, 1,1'-biphenyl-4-sulfonylchloride, biphenyl-4-sulphonyl chloride, 4-phenylbenzene-1-sulfonyl chloride, 1,1-biphenyl-4-sulfonylchloride, pubchem20990 |
PubChem CID | 74192 |
SMILES | ClS(=O)(=O)C1=CC=C(C=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11470117
|
Thermo Scientific Alfa Aesar
B21031.06 |
5 g |
59.50 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11480117
|
Thermo Scientific Alfa Aesar
B21031.14 |
25 g |
188.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11490117
|
Thermo Scientific Alfa Aesar
B21031.22 |
100 g |
690.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is a reactant for preparation of anilinopyrimidinesulfonamides as potential anticancer agents, synthesis of inhibitors of the hypoxia inducible factor pathway. It participates in the preparation of hydroxy-sulfonyl-piperidinyl butyramides as HDAC inhibitors and potential antitumors and is also involved in palladium-catalyzed desulfitative C-arylation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a reactant for preparation of anilinopyrimidinesulfonamides as potential anticancer agents, synthesis of inhibitors of the hypoxia inducible factor pathway. It participates in the preparation of hydroxy-sulfonyl-piperidinyl butyramides as HDAC inhibitors and potential antitumors and is also involved in palladium-catalyzed desulfitative C-arylation.
Solubility
Reacts with water.
Notes
Store at room temperature. Keep away from moisture, bases and oxidizing agents.
Chemical Identifiers
1623-93-4 | |
252.71 | |
ALBQXDHCMLLQMB-UHFFFAOYSA-N | |
74192 |
C12H9ClO2S | |
MFCD00078658 | |
4-biphenylsulfonyl chloride, 1,1'-biphenyl-4-sulfonyl chloride, biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulphonyl chloride, 4-biphenylsulfonylchloride, 1,1'-biphenyl-4-sulfonylchloride, biphenyl-4-sulphonyl chloride, 4-phenylbenzene-1-sulfonyl chloride, 1,1-biphenyl-4-sulfonylchloride, pubchem20990 | |
ClS(=O)(=O)C1=CC=C(C=C1)C1=CC=CC=C1 |
Specifications
1623-93-4 | |
200°C to 205°C (6 mmHg) | |
MFCD00078658 | |
2694373 | |
4-biphenylsulfonyl chloride, 1,1'-biphenyl-4-sulfonyl chloride, biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulphonyl chloride, 4-biphenylsulfonylchloride, 1,1'-biphenyl-4-sulfonylchloride, biphenyl-4-sulphonyl chloride, 4-phenylbenzene-1-sulfonyl chloride, 1,1-biphenyl-4-sulfonylchloride, pubchem20990 | |
ALBQXDHCMLLQMB-UHFFFAOYSA-N | |
4-phenylbenzenesulfonyl chloride | |
74192 | |
97% |
112°C to 117°C | |
C12H9ClO2S | |
UN3261 | |
Moisture sensitive | |
Reacts with water. | |
ClS(=O)(=O)C1=CC=C(C=C1)C1=CC=CC=C1 | |
252.71 | |
252.72 | |
Biphenyl-4-sulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 216-609-1
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.