missing translation for 'onlineSavingsMsg'
Learn More

Nitritex™ BioClean Nano 5™ Non-sterile Nitrile Gloves, ISO Class 5

Product Code. 15278268
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

With textured fingers to provide enhanced grip. Nitritex™ BioClean Nano 5™ Non-sterile Nitrile Gloves, ISO Class 5 are antistatic, powder-free, latex-free, resistant to a wide range of chemicals and are ideal for use in electrically sensitive applications.

  • Resistant to a wide range of chemicals
  • Textured fingers for enhanced grip
  • Powder-free and latex-free
  • Excellent antistatic properties
  • Non-particulating EasyTear™ packaging
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) ISO 9001:2015 and (EU)2016/425, EN420:2003 + A1:2009, EN374-1:20016, EN374-5:2016 and EN421:2010, EN455-2:2015, (EC)1935/2004 and (EC)2023/2006
Color White
Disposable Single-use
Exterior Finish Fully Textured
Grade Category 3 - PPE Directive 89/686/EEC
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size 2X-Large
Sterility Non-sterile
Finger Style Textured
For Use With (Application) Electrically sensitivity applications
Glove Cuff Style Beaded
Packaging Type 100 pieces per sealed inner PE bag; one inner PE bag per sealed outer PE bag; 10 outer bags per lined carton (1000 pieces)
Quantity 100 pieces per sealed inner PE bag; one inner PE bag per sealed outer PE bag; 10 outer bags per lined carton (1000 pieces)
Resistant To A broad range of chemicals
Series BioClean Nano 5
Length (English) 12 in.
Length (Metric) 30 cm
Width (Metric) 119 mm ± 5
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'