missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell BioClean-D™ S-BDSH Drop-down Sterile Coverall with Hood
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Description
- Sterile cleanroom coverall with hood features drop-down design technology, preventing needless external surface contact.
- Equipped with quick-release tabs and internal colored tabs, indicating safe touch points that further ensure aseptic donning.
- Elastic back, hood, ankles, cuffs.
- Thumb loops to ensure a secure hold and fit.
- Antistatic garment is manufactured from low-linting CleanTough™ material to reduce particle contamination risks.

Specifications
Specifications
Product Type | Cleanroom Coveralls |
Size | Medium |
Gender | Unisex |
Material | Polypropylene |
Color | White |
Disposable | Single-use |
Waist Style | Standard |
Ankle Style | Elastic |
Cuff Style | Elastic |
Neck Style | Hooded |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ansell BioClean-D™ S-BDSH Drop-down Sterile Coverall with Hood >
Spot an opportunity for improvement?Share a Content Correction