missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BICINE, 0.5M buffer soln., pH 9.0, Thermo Scientific™
52.80 € - 82.10 €
Chemical Identifiers
CAS | 150-25-4 |
---|---|
Molecular Formula | C6H12NNaO4 |
Molecular Weight (g/mol) | 185.16 |
MDL Number | MFCD00004295 |
InChI Key | MFBDBXAVPLFMNJ-UHFFFAOYSA-M |
PubChem CID | 8761 |
ChEBI | CHEBI:40957 |
IUPAC Name | 2-[bis(2-hydroxyethyl)amino]acetic acid |
SMILES | [Na+].OCCN(CCO)CC([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15415149
|
Thermo Scientific Alfa Aesar
J62838.AE |
100 mL |
52.80 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15425149
|
Thermo Scientific Alfa Aesar
J62838.AK |
250 mL |
82.10 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemische Identifikatoren
150-25-4 | |
185.16 | |
MFBDBXAVPLFMNJ-UHFFFAOYSA-M | |
CHEBI:40957 | |
[Na+].OCCN(CCO)CC([O-])=O |
C6H12NNaO4 | |
MFCD00004295 | |
8761 | |
2-[bis(2-hydroxyethyl)amino]acetic acid |
Spezifikation
150-25-4 | |
9.0 | |
C6H12NNaO4 | |
MFBDBXAVPLFMNJ-UHFFFAOYSA-M | |
2-[bis(2-hydroxyethyl)amino]acetic acid | |
8761 | |
BICINE buffer soln. |
0.5 M | |
Liquid | |
MFCD00004295 | |
[Na+].OCCN(CCO)CC([O-])=O | |
185.16 | |
CHEBI:40957 |
Sicherheit und Handhabung
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures