missing translation for 'onlineSavingsMsg'
Learn More

beta-Propiolactone, 98%, pure, Thermo Scientific Chemicals

Chemical Identifiers

CAS 57-57-8
Molecular Formula C3H4O2
Molecular Weight (g/mol) 72.06
MDL Number MFCD00005169
InChI Key VEZXCJBBBCKRPI-UHFFFAOYSA-N
Synonym beta-propiolactone, propiolactone, 2-oxetanone, propanolide, betaprone, 3-propanolide, 1,3-propiolactone, 3-propiolactone, beta-propionolactone, beta-propiolakton
PubChem CID 2365
ChEBI CHEBI:49073
IUPAC Name oxetan-2-one
SMILES O=C1CCO1
View More Specs

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

57-57-8
72.06
VEZXCJBBBCKRPI-UHFFFAOYSA-N
2365
oxetan-2-one
C3H4O2
MFCD00005169
beta-propiolactone, propiolactone, 2-oxetanone, propanolide, betaprone, 3-propanolide, 1,3-propiolactone, 3-propiolactone, beta-propionolactone, beta-propiolakton
CHEBI:49073
O=C1CCO1

Specifications

57-57-8
98%
MFCD00005169
VEZXCJBBBCKRPI-UHFFFAOYSA-N
oxetan-2-one
2365
72.06
β-Propiolactone, Pure
1.1460g/mL
C3H4O2
beta-propiolactone, propiolactone, 2-oxetanone, propanolide, betaprone, 3-propanolide, 1,3-propiolactone, 3-propiolactone, beta-propionolactone, beta-propiolakton
O=C1CCO1
72.06
CHEBI:49073
Pure
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Fatal if inhaled.
May cause cancer.

GHS P Statement:
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only