missing translation for 'onlineSavingsMsg'
Learn More

beta-Nicotinamide adenine dinucleotide phosphate monosodium salt, Thermo Scientific Chemicals

145.73 € - 412.05 €

Chemical Identifiers

CAS 1184-16-3
Molecular Formula C21H27N7NaO17P3
Molecular Weight (g/mol) 765.39
MDL Number MFCD00036973
InChI Key JNUMDLCHLVUHFS-QYZPTAICSA-M
Synonym nadp sodium salt, beta-nicotinamide adenine dinucleotide phosphate sodium
PubChem CID 131845262
IUPAC Name [[(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate;sodium
SMILES [Na+].NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP(O)(O)=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11339608
View Documents
Thermo Scientific Alfa Aesar
044126.MD
250 mg
145.73 €
250mg
Please to purchase this item. Need a web account? Register with us today!
11349608
View Documents
Thermo Scientific Alfa Aesar
044126.03
1 g
412.05 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

β-Nicotinamide adenine dinucleotide phosphate monosodium salt acts as an electron acceptor used by cells. It is also useful in vitro to study NADP. Furthermore, it is used in a wide range of enzyme catalyzed oxidation reduction reactions. The combination of NADP+/NADPH is employed in a variety of antioxidation mechanisms, which protect against reactive oxidation species accumulation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1184-16-3
765.39
JNUMDLCHLVUHFS-QYZPTAICSA-M
131845262
[Na+].NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP(O)(O)=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
C21H27N7NaO17P3
MFCD00036973
nadp sodium salt, beta-nicotinamide adenine dinucleotide phosphate sodium
[[(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate;sodium

Specifications

1184-16-3
C21H27N7NaO17P3
4779954
Soluble in water.
[Na+].NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP(O)(O)=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
765.39
765.4
beta-Nicotinamide adenine dinucleotide phosphate monosodium salt
White
MFCD00036973
nadp sodium salt, beta-nicotinamide adenine dinucleotide phosphate sodium
JNUMDLCHLVUHFS-QYZPTAICSA-M
[[(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate;sodium
131845262
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'einecsNumber' : 214-664-6

missing translation for 'tsca' : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.