Learn More
beta-Nicotinamide adenine dinucleotide, 97%, Thermo Scientific Chemicals
49.70 € - 722.00 €
Chemical Identifiers
CAS | 53-84-9 |
---|---|
Molecular Formula | C21H27N7O14P2 |
Molecular Weight (g/mol) | 663.43 |
MDL Number | MFCD00150377 |
InChI Key | BAWFJGJZGIEFAR-WIWLTUSXNA-N |
Synonym | nicotinamide adenine dinucleotide, nad+, diphosphopyridine nucleotide, nad-oxidized, nicotinamide-adenine dinucleotide, dpn-ox, beta-nicotinamide adenine dinucleotide, dpn+, nad, nad + |
PubChem CID | 15938971 |
ChEBI | CHEBI:57540 |
IUPAC Name | [[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate |
SMILES | NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15424739
|
Thermo Scientific Alfa Aesar
J62337.03 |
1 g |
49.70 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15434739
|
Thermo Scientific Alfa Aesar
J62337.06 |
5 g |
198.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15444739
|
Thermo Scientific Alfa Aesar
J62337.14 |
25 g |
722.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NAD+, Free Acid is a major electron acceptor molecule in biological oxidation reactions. It is also used as a catalyst intermediate and several enzymes in a multistep system, known as enzyme cycling,.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNAD+, Free Acid is a major electron acceptor molecule in biological oxidation reactions. It is also used as a catalyst intermediate and several enzymes in a multistep system, known as enzyme cycling,.
Solubility
Soluble in water at 50mg/ml
Notes
b-NAD is very hygroscopic and should be stored desiccated. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C.
Chemical Identifiers
53-84-9 | |
663.43 | |
BAWFJGJZGIEFAR-WIWLTUSXNA-N | |
15938971 | |
[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate |
C21H27N7O14P2 | |
MFCD00150377 | |
nicotinamide adenine dinucleotide, nad+, diphosphopyridine nucleotide, nad-oxidized, nicotinamide-adenine dinucleotide, dpn-ox, beta-nicotinamide adenine dinucleotide, dpn+, nad, nad + | |
CHEBI:57540 | |
NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O |
Specifications
53-84-9 | |
C21H27N7O14P2 | |
3584133 | |
nicotinamide adenine dinucleotide, nad+, diphosphopyridine nucleotide, nad-oxidized, nicotinamide-adenine dinucleotide, dpn-ox, beta-nicotinamide adenine dinucleotide, dpn+, nad, nad + | |
BAWFJGJZGIEFAR-WIWLTUSXNA-N | |
[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate | |
15938971 | |
663.43 | |
Powder |
White | |
MFCD00150377 | |
14,6344 | |
Soluble in water at 50mg/ml | |
NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O | |
663.43 | |
CHEBI:57540 | |
≥98% | |
beta-Nicotinamide adenine dinucleotide |
Safety and Handling
missing translation for 'einecsNumber' : 200-184-4
missing translation for 'rtecsNumber' : UU3450000
missing translation for 'tsca' : Yes
Recommended Storage : Store at -20°C
RUO – Research Use Only