Learn More
Beta Actin Mouse anti-Human, HRP, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech HRP-60008-100UL
Product Code. 16801246
  • 189.00 € / 100µL
Explore more special offers
Description
Description
The Beta Actin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Beta Actin. This antibody recognizes human, mouse, rat, zebrafish, plant antigen. The Beta Actin antibody has been validated for the following applications: WB analysis. This antibody is hrp.
Specifications
Specifications
Beta Actin
Monoclonal
HRP
100 μL
60
IgM
Western Blot, Immunofluorescence, ELISA
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Zebrafish, Plant