missing translation for 'onlineSavingsMsg'
Learn More

Beryllium wire, 0.25mm (0.01 in.) dia., annealed, 99.7% (metals basis), Thermo Scientific Chemicals

859.00 € - 1554.00 €

Chemical Identifiers

CAS 7440-41-7
Molecular Formula Be
Molecular Weight (g/mol) 9.01
MDL Number MFCD00134032
InChI Key ATBAMAFKBVZNFJ-UHFFFAOYSA-N
Synonym glucinium, dust, metal, powder, metallic, berilio, beryllium, elemental, glucinum, beryllium-9, and compounds
PubChem CID 5460467
ChEBI CHEBI:33783
IUPAC Name beryllium
SMILES [Be]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11358618
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040680.BU
25 cm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11368618
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040680.BW
50 cm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-41-7
9.01
ATBAMAFKBVZNFJ-UHFFFAOYSA-N
5460467
beryllium
Be
MFCD00134032
glucinium, dust, metal, powder, metallic, berilio, beryllium, elemental, glucinum, beryllium-9, and compounds
CHEBI:33783
[Be]

Specifications

7440-41-7
MFCD00134032
ATBAMAFKBVZNFJ-UHFFFAOYSA-N
beryllium
5460467
99.7%
≈0.091g/m
(metals basis)
Be
glucinium, dust, metal, powder, metallic, berilio, beryllium, elemental, glucinum, beryllium-9, and compounds
[Be]
9.01
CHEBI:33783
Odorless
Wire
Beryllium wire, 0.25mm (0.01 in.) dia., Annealed, Temper: soft
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H330-H350-H372-H315-H319-H317-H335
Toxic if swallowed.
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

P201-P202-P260-P264b-P270-P271-P272-P280-P281-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P362-P501c

H301-H315-H317-H319-H330-H335-H350i-H372

missing translation for 'einecsNumber' : 231-150-7

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only