Learn More
Benzyltrimethylammonium hydroxide, 40 wt% solution in methanol, Thermo Scientific Chemicals
115.00 € - 324.00 €
Chemical Identifiers
CAS | 100-85-6, 67-56-1 |
---|---|
Molecular Formula | C10H17NO |
Molecular Weight (g/mol) | 167.252 |
MDL Number | MFCD00008281 |
InChI Key | NDKBVBUGCNGSJJ-UHFFFAOYSA-M |
Synonym | benzyltrimethylammonium hydroxide, triton b, n,n,n-trimethyl-1-phenylmethanaminium hydroxide, trimethylbenzylammonium hydroxide, benzyl trimethylammonium hydroxide, benzyl trimethyl ammonium hydroxide, trimethyl benzylammonium hydroxide, sumquat 2311, benzyltrimetylammonium hydroxide, n,n,n-trimethylbenzenemethanaminium hydroxide |
PubChem CID | 66854 |
IUPAC Name | benzyl(trimethyl)azanium;hydroxide |
SMILES | C[N+](C)(C)CC1=CC=CC=C1.[OH-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10519810
|
Thermo Scientific Acros
106200010 |
1 L | Glass bottle |
115.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10425565
|
Thermo Scientific Acros
106200051 |
5 L | Plastic Drum |
324.00 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystChemical Identifiers
100-85-6, 67-56-1 | |
167.252 | |
NDKBVBUGCNGSJJ-UHFFFAOYSA-M | |
66854 | |
C[N+](C)(C)CC1=CC=CC=C1.[OH-] |
C10H17NO | |
MFCD00008281 | |
benzyltrimethylammonium hydroxide, triton b, n,n,n-trimethyl-1-phenylmethanaminium hydroxide, trimethylbenzylammonium hydroxide, benzyl trimethylammonium hydroxide, benzyl trimethyl ammonium hydroxide, trimethyl benzylammonium hydroxide, sumquat 2311, benzyltrimetylammonium hydroxide, n,n,n-trimethylbenzenemethanaminium hydroxide | |
benzyl(trimethyl)azanium;hydroxide |
Specifications
Yellow | |
39.0, 59.0 | |
C10H17NO | |
MFCD00008281 | |
0.9250g/mL | |
Solubility in water: miscible. | |
C[N+](C)(C)CC1=CC=CC=C1.[OH-] | |
167.252 | |
167.25 | |
0.925 | |
Solution |
100-85-6, 67-56-1 | |
41.0, 61.0 | |
C6H5CH2N(CH3)3OH | |
01,1252; 05,29; 08,36; 12,48; 05,30 | |
benzyltrimethylammonium hydroxide, triton b, n,n,n-trimethyl-1-phenylmethanaminium hydroxide, trimethylbenzylammonium hydroxide, benzyl trimethylammonium hydroxide, benzyl trimethyl ammonium hydroxide, trimethyl benzylammonium hydroxide, sumquat 2311, benzyltrimetylammonium hydroxide, n,n,n-trimethylbenzenemethanaminium hydroxide | |
NDKBVBUGCNGSJJ-UHFFFAOYSA-M | |
benzyl(trimethyl)azanium;hydroxide | |
66854 | |
≥40% | |
15°C | |
Benzyltrimethylammonium hydroxide, 40 wt. %solution in Methanol |
Safety and Handling
GHS H Statement:
Toxic if inhaled.
Toxic in contact with skin.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes damage to organs.
Highly flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
missing translation for 'einecsNumber' : 202-895-5
missing translation for 'rtecsNumber' : BO8575000
missing translation for 'tsca' : TSCA
- Benzyltrimethylammonium hydroxide, 40 wt% solution in methanol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H370-Causes damage to organs.
- H314-Causes severe skin burns and eye damage.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only