missing translation for 'onlineSavingsMsg'
Learn More

Benzyl glycinate hydrochloride, 98%, Thermo Scientific Chemicals

36.20 € - 127.00 €

Chemical Identifiers

CAS 2462-31-9
Molecular Formula C9H12ClNO2
Molecular Weight (g/mol) 201.65
MDL Number MFCD00035442
InChI Key VLQHNAMRWPQWNK-UHFFFAOYSA-N
Synonym benzyl glycinate hydrochloride, benzyl 2-aminoacetate hydrochloride, glycine benzyl ester hydrochloride, h-dl-gly-obzl.hcl, benzyl glycinate hcl, h-gly-obzl.hcl, benzyl glycinate, h-gly-obzl hcl, glycine benzyl ester hcl, glycine, phenylmethyl ester, hydrochloride
PubChem CID 11701227
IUPAC Name benzyl 2-aminoacetate;hydrochloride
SMILES C1=CC=C(C=C1)COC(=O)CN.Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11459370
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H50247.03
1 g
36.20 €
1g
Please to purchase this item. Need a web account? Register with us today!
11469370
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H50247.06
5 g
127.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2462-31-9
201.65
VLQHNAMRWPQWNK-UHFFFAOYSA-N
11701227
C1=CC=C(C=C1)COC(=O)CN.Cl
C9H12ClNO2
MFCD00035442
benzyl glycinate hydrochloride, benzyl 2-aminoacetate hydrochloride, glycine benzyl ester hydrochloride, h-dl-gly-obzl.hcl, benzyl glycinate hcl, h-gly-obzl.hcl, benzyl glycinate, h-gly-obzl hcl, glycine benzyl ester hcl, glycine, phenylmethyl ester, hydrochloride
benzyl 2-aminoacetate;hydrochloride

Specifications

2462-31-9
C9H12ClNO2
MFCD00035442
VLQHNAMRWPQWNK-UHFFFAOYSA-N
benzyl 2-aminoacetate;hydrochloride
11701227
98%
139°C to 140°C
H2NCH2CO2CH2C6H5·HCl
benzyl glycinate hydrochloride, benzyl 2-aminoacetate hydrochloride, glycine benzyl ester hydrochloride, h-dl-gly-obzl.hcl, benzyl glycinate hcl, h-gly-obzl.hcl, benzyl glycinate, h-gly-obzl hcl, glycine benzyl ester hcl, glycine, phenylmethyl ester, hydrochloride
C1=CC=C(C=C1)COC(=O)CN.Cl
201.65
201.65
Benzyl glycinate hydrochloride
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only