missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 4-hydroxyphenyl ketone, 97%, Thermo Scientific Chemicals

Product Code. 10421094
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2491-32-9
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00002360
InChI Key JBQTZLNCDIFCCO-UHFFFAOYSA-N
Synonym benzyl 4-hydroxyphenyl ketone, 1-4-hydroxyphenyl-2-phenylethanone, 4-hydroxydeoxybenzoin, acetophenone, 4'-hydroxy-2-phenyl, 1-4-hydroxyphenyl-2-phenylethan-1-one, ethanone, 1-4-hydroxyphenyl-2-phenyl, acmc-20alpm, 4-hydroxydesoxybenzoin, p-phenylacetyl phenol, maybridge3_000426
PubChem CID 75607
IUPAC Name 1-(4-hydroxyphenyl)-2-phenylethanone
SMILES C1=CC=C(C=C1)CC(=O)C2=CC=C(C=C2)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 148°C to 151°C
Assay Percent Range 96% min. (HPLC)
Linear Formula C6H5CH2C(O)C6H4OH
Quantity 25 g
Beilstein 08, 165
Solubility Information Solubility in water: insoluble
Formula Weight 212.25
Percent Purity 97%
Chemical Name or Material Benzyl 4-hydroxyphenyl ketone
Show More Show Less
compliance-icons
Product Identifier
  • Benzyl 4-hydroxyphenyl ketone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.