missing translation for 'onlineSavingsMsg'
Learn More

Benzoic Acid Melting Point Standard, Reagecon™
BIOPHARMA

Product Code. 13592100
Click to view available options
Quantity:
1 g
Packaging:
1g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Packaging 1g
Quantity 1 g
compliance-icons compliance-icons
Product Identifier
  • MELTING POINT STANDARD BENZOIC ACID 121 to 123°C
Signal Word
  • Danger
Hazard Category
  • Specific target organ toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'