Learn More
Benzoic acid, 99.6%, ACS reagent, Thermo Scientific Chemicals
21.31 € - 835.16 €
Chemical Identifiers
CAS | 65-85-0 |
---|---|
Molecular Formula | C7H6O2 |
Molecular Weight (g/mol) | 122.12 |
InChI Key | WPYMKLBDIGXBTP-UHFFFAOYSA-N |
Synonym | benzenecarboxylic acid, dracylic acid, benzeneformic acid, carboxybenzene, phenylformic acid, benzenemethanoic acid, phenylcarboxylic acid, retardex, benzoesaeure gk, benzoesaeure gv |
PubChem CID | 243 |
ChEBI | CHEBI:30746 |
IUPAC Name | benzoic acid |
SMILES | C1=CC=C(C=C1)C(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10627231
|
Thermo Scientific Acros
423470250 |
25 g | Plastic bottle |
21.31 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10005860
|
Thermo Scientific Acros
423475000 |
500 g | Plastic bottle |
85.63 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10799911
|
Thermo Scientific Acros
423470020 |
2 kg | Plastic bottle |
216.08 €
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11974971
|
Thermo Scientific Acros
423470100 |
10 kg | Plastic drum |
835.16 €
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
65-85-0 | |
122.12 | |
benzenecarboxylic acid, dracylic acid, benzeneformic acid, carboxybenzene, phenylformic acid, benzenemethanoic acid, phenylcarboxylic acid, retardex, benzoesaeure gk, benzoesaeure gv | |
CHEBI:30746 | |
C1=CC=C(C=C1)C(=O)O |
C7H6O2 | |
WPYMKLBDIGXBTP-UHFFFAOYSA-N | |
243 | |
benzoic acid |
Specifications
65-85-0 | |
100.0 | |
White | |
121°C | |
Authentic | |
C7H6O2 | |
09,92 | |
15,1093 | |
Solubility in water: 3g/L (20°C). Other solubilities: soluble in alcohol,benzene,ether,chloroform,,ccl4,carbon disulfide and turpentine | |
C1=CC=C(C=C1)C(=O)O | |
122.12 | |
CHEBI:30746 | |
99.6% | |
0.005% max. (in methanol) | |
Benzoic acid |
99.5 | |
121.0°C to 123.0°C | |
249.0°C | |
0.005% max. | |
99.6% | |
C6H5CO2H | |
01,49; 13,312 | |
benzenecarboxylic acid, dracylic acid, benzeneformic acid, carboxybenzene, phenylformic acid, benzenemethanoic acid, phenylcarboxylic acid, retardex, benzoesaeure gk, benzoesaeure gv | |
WPYMKLBDIGXBTP-UHFFFAOYSA-N | |
benzoic acid | |
243 | |
122.12 | |
ACS Reagent | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye damage.
Causes damage to organs through prolonged or repeated exposure if inhaled.
GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for s
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-618-2
- Benzoic acid
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 1
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H372-Causes damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.