missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzo[b]thiophene 1,1-dioxide, 98%, Thermo Scientific Chemicals
120.00 € - 439.00 €
Chemical Identifiers
CAS | 825-44-5 |
---|---|
Molecular Formula | C8H6O2S |
Molecular Weight (g/mol) | 166.19 |
MDL Number | MFCD00052178 |
InChI Key | FRJNKYGTHPUSJR-UHFFFAOYSA-N |
Synonym | benzo b thiophene 1,1-dioxide, benzothiophene 1,1-dioxide, thianaphthene 1,1-dioxide, benzo b thiophene, 1,1-dioxide, thianaphthene-1,1-dioxide, benzo b thiophene s,s-dioxide, benzo b thiole-1,1-dione, benzothiophene sulfone, maybridge1_005454 |
PubChem CID | 69997 |
IUPAC Name | 1-benzothiophene 1,1-dioxide |
SMILES | O=S1(=O)C=CC2=CC=CC=C12 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
11477986
|
Thermo Scientific Alfa Aesar
A14063.06 |
5 g |
120.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11487986
|
Thermo Scientific Alfa Aesar
A14063.14 |
25 g |
439.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemische Identifikatoren
825-44-5 | |
166.19 | |
FRJNKYGTHPUSJR-UHFFFAOYSA-N | |
69997 | |
O=S1(=O)C=CC2=CC=CC=C12 |
C8H6O2S | |
MFCD00052178 | |
benzo b thiophene 1,1-dioxide, benzothiophene 1,1-dioxide, thianaphthene 1,1-dioxide, benzo b thiophene, 1,1-dioxide, thianaphthene-1,1-dioxide, benzo b thiophene s,s-dioxide, benzo b thiole-1,1-dione, benzothiophene sulfone, maybridge1_005454 | |
1-benzothiophene 1,1-dioxide |
Spezifikation
825-44-5 | |
C8H6O2S | |
116442 | |
FRJNKYGTHPUSJR-UHFFFAOYSA-N | |
1-benzothiophene 1,1-dioxide | |
69997 | |
98% |
137°C to 138°C | |
MFCD00052178 | |
benzo b thiophene 1,1-dioxide, benzothiophene 1,1-dioxide, thianaphthene 1,1-dioxide, benzo b thiophene, 1,1-dioxide, thianaphthene-1,1-dioxide, benzo b thiophene s,s-dioxide, benzo b thiole-1,1-dione, benzothiophene sulfone, maybridge1_005454 | |
O=S1(=O)C=CC2=CC=CC=C12 | |
166.19 | |
166.2 | |
Benzo[b]thiophene 1,1-dioxide |
Sicherheit und Handhabung
missing translation for 'einecsNumber' : 212-544-8
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only