missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]furan, 97+%, Thermo Scientific Chemicals

58.09 € - 255.27 €

Chemical Identifiers

CAS 271-89-6
Molecular Formula C8H6O
Molecular Weight (g/mol) 118.14
MDL Number MFCD00005847
InChI Key IANQTJSKSUMEQM-UHFFFAOYSA-N
Synonym benzofuran, 2,3-benzofuran, coumarone, benzo b furan, cumarone, benzofurfuran, 1-oxindene, coumaron, 1-oxidene
PubChem CID 9223
ChEBI CHEBI:35260
IUPAC Name 1-benzofuran
SMILES O1C=CC2=CC=CC=C12
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11432258
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A15225.09
10 g
58.09 €
10g
Please to purchase this item. Need a web account? Register with us today!
11442258
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A15225.18
50 g
255.27 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Benzo[b]furan is used in the manufacture of coumarone-indene resin. It has been used to make floor tiles and other products. It is also used as a coating on grapefruit, lemons, lirnes, oranges, tangelos and tangerines and in the production of COITosion-resistant paints and varnishes, in water-resistant coatings on paper products and fabrics and as adhesives in food containers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzo[b]furan is used in the manufacture of coumarone-indene resin. It has been used to make floor tiles and other products. It is also used as a coating on grapefruit, lemons, lirnes, oranges, tangelos and tangerines and in the production of COITosion-resistant paints and varnishes, in water-resistant coatings on paper products and fabrics and as adhesives in food containers.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Specifications

Chemical Identifiers

271-89-6
118.14
IANQTJSKSUMEQM-UHFFFAOYSA-N
9223
1-benzofuran
C8H6O
MFCD00005847
benzofuran, 2,3-benzofuran, coumarone, benzo b furan, cumarone, benzofurfuran, 1-oxindene, coumaron, 1-oxidene
CHEBI:35260
O1C=CC2=CC=CC=C12

Specifications

271-89-6
173°C to 175°C
Aromatic
1.566
UN1993
14,1088
Not miscible in water.
O1C=CC2=CC=CC=C12
118.14
CHEBI:35260
≥99%
1.095
56°C (133°F)
C8H6O
MFCD00005847
107704
benzofuran, 2,3-benzofuran, coumarone, benzo b furan, cumarone, benzofurfuran, 1-oxindene, coumaron, 1-oxidene
IANQTJSKSUMEQM-UHFFFAOYSA-N
1-benzofuran
9223
118.14
Benzo[b]furan
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H351
Flammable liquid and vapour.
Suspected of causing cancer.

P201-P202-P210-P233-P240-P241-P242-P243-P281-P303+P361+P353-P308+P313-P370+P378q-P501c

H226-H351

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

missing translation for 'einecsNumber' : 205-982-6

missing translation for 'rtecsNumber' : DF6423800

missing translation for 'tsca' : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only