missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]furan-3-boronic acid, 98%, Thermo Scientific Chemicals

Product Code. 15461579
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 317830-83-4
Molecular Formula C8H7BO3
Molecular Weight (g/mol) 161.95
MDL Number MFCD06801687
InChI Key DFUGYZQSDFQVPU-UHFFFAOYSA-N
Synonym benzofuran-3-ylboronic acid, benzofuran-3-boronic acid, 1-benzofuran-3-yl boronic acid, boronic acid, 3-benzofuranyl, acmc-1csxn, benzo b furan-3-boronic acid, benzofuran-3-yl boronic acid
PubChem CID 23438904
SMILES OB(O)C1=COC2=CC=CC=C12

Specifications

Quantity 5 g
Beilstein 9479377
IUPAC Name 1-benzofuran-3-ylboronic acid
Formula Weight 161.95
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Benzo[b]furan-3-boronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.