missing translation for 'onlineSavingsMsg'
Learn More

Benzil, 99+%, Thermo Scientific Chemicals

32.86 € - 222.11 €

Chemical Identifiers

CAS 134-81-6
Molecular Formula C14H10O2
Molecular Weight (g/mol) 210.23
MDL Number MFCD00003080
InChI Key WURBFLDFSFBTLW-UHFFFAOYSA-N
Synonym benzil, diphenylethanedione, dibenzoyl, diphenylglyoxal, bibenzoyl, ethanedione, diphenyl, 1,2-diphenylethanedione, diphenylethane-1,2-dione, glyoxal, diphenyl, diphenyldiketon
PubChem CID 8651
ChEBI CHEBI:51507
IUPAC Name 1,2-diphenylethane-1,2-dione
SMILES O=C(C(=O)C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10346070
View Documents
Thermo Scientific Acros
105411000
100 g Plastic bottle
32.86 €
100g
Please to purchase this item. Need a web account? Register with us today!
10677052
View Documents
Thermo Scientific Acros
105415000
500 g Plastic bottle
70.65 €
500g
Please to purchase this item. Need a web account? Register with us today!
11964841
View Documents
Thermo Scientific Acros
105410025
2.5 kg Plastic bottle
222.11 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

134-81-6
210.23
WURBFLDFSFBTLW-UHFFFAOYSA-N
8651
1,2-diphenylethane-1,2-dione
C14H10O2
MFCD00003080
benzil, diphenylethanedione, dibenzoyl, diphenylglyoxal, bibenzoyl, ethanedione, diphenyl, 1,2-diphenylethanedione, diphenylethane-1,2-dione, glyoxal, diphenyl, diphenyldiketon
CHEBI:51507
O=C(C(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

134-81-6
100.0
White
0.5% max.
99% min. (GC)
C6H5COCOC6H5
15,108
Solubility in water: 0.5g/L (20°C). Other solubilities: soluble in alcohol,ether,chloroform,ethyl,acetate,benzene,toluene and nitrobenzene
O=C(C(=O)C1=CC=CC=C1)C1=CC=CC=C1
210.23
CHEBI:51507
99+%
Benzil, 99+%
99.0
94.0°C to 97.0°C
346.0°C to 348.0°C
Authentic
C14H10O2
MFCD00003080
benzil, diphenylethanedione, dibenzoyl, diphenylglyoxal, bibenzoyl, ethanedione, diphenyl, 1,2-diphenylethanedione, diphenylethane-1,2-dione, glyoxal, diphenyl, diphenyldiketon
WURBFLDFSFBTLW-UHFFFAOYSA-N
1,2-diphenylethane-1,2-dione
8651
210.23
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 205-157-0

missing translation for 'rtecsNumber' : DD1925000

missing translation for 'tsca' : TSCA

Product Identifier
  • Benzil

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.