missing translation for 'onlineSavingsMsg'
Learn More

Benzalkonium chloride, MP Biomedicals™

58.80 € - 123.00 €

Chemical Identifiers

CAS 8001-54-5
Molecular Formula C9H13ClN
Molecular Weight (g/mol) 170.66
Synonym Alkyldimethylbenzyl Ammonium Chloride, Ammonyx
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17851421
View Documents Promotion Details
MP Biomedicals
0215043083
250 g
58.80 €
250g
Please to purchase this item. Need a web account? Register with us today!
17861421
View Documents Promotion Details
MP Biomedicals
0215043091
1 kg
123.00 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

8001-54-5
C9H13ClN
170.66
Alkyldimethylbenzyl Ammonium Chloride, Ammonyx

Specifications

Colorless
Viscous Solution
Alkyldimethylbenzyl Ammonium Chloride, Ammonyx
Benzalkonium chloride
8001-54-5
C9H13ClN
170.66
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at Room Temperature(15°C to 30°C).

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

SDS
Documents

Documents

Product Certifications