missing translation for 'onlineSavingsMsg'
Learn More

Benzaldehyde, 99+%, Extra Pure, SLR, Fisher Chemical

Product Code. 10050020
Click to view available options
500mL, Plastic Coated Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 100-52-7
Melting Point -26°C
pH 5.9
Boiling Point 179°C
Packaging Plastic Coated Glass bottle
Molecular Formula C7H6O
MDL Number 3299
Quantity 500 mL
Formula Weight 106.12g/mol
Physical Form Liquid
compliance-icons
Product Identifier
  • Benzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /A§
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.